
2,4,5,6-Tetraaminopyrimidine sulphate ≥98.0% (by titrimetric analysis)
Catalog # TCIAT1462-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (HPLC):min. 95.0 area%
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
About this item
:
:238.23 g/mol
EINECS:226-393-0
:GHS07
CAS Number:5392-28-9
Formula:C₄H₈N₆·H₂SO₄
MDL Number:MFCD00012786