
2,4,5,6-Tetraaminopyrimidine sulphate hydrate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5392-28-9
:
Formula:C₄H₈N₆·H₂SO₄·xH₂O
MDL Number:MFCD06408008
:238.23 g/mol (anhy)
Melting Pt:>300 °C
EINECS:226-393-0
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Ochre to brown Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤9 % (1 g, 150°C, 3 hrs)
- Titration with NaOH:≥97.5 % (On dry substance)
Specification Test Results
Frequently Bought Together