N-Acetyl-5-hydroxytryptamine ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:1210-83-9
:
Formula:C₁₂H₁₄N₂O₂
MDL Number:MFCD00005656
:218.3 g/mol
EINECS:214-916-5
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:119.0 to 122.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together