Order Entry
Italy
ContactUsLinkComponent
O-Methylhydroxylamine hydrochloride ∼40% ∼5.4 mol/L in water
Corrosive
Irritant
Environment
O-Methylhydroxylamine hydrochloride ∼40% ∼5.4 mol/L in water
Supplier:  TCI
Certificates
undefined
Corrosive
Irritant
Environment
O-Methylhydroxylamine hydrochloride ∼40% ∼5.4 mol/L in water
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Concentration (Neutralization titration):
    40.0 to 44.0 w/w%

Specification Test Results

Frequently Bought Together