
1-Bromo-2-(trifluoromethylthio)benzene ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1644-72-0
:
Formula:C₇H₄BrF₃S
MDL Number:MFCD00236347
:257.07 g/mol
Boiling Pt:92…94 °C (20 mmHg)
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together