
2'-Methylacetoacetanilide ≥98.0% (by HPLC, total nitrogen)
: TCI
:93-68-5
:
:C₁₁H₁₃NO₂
:MFCD00008782
:191.23 g/mol
:202-267-0
:GHS07
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:104.0 to 108.0 deg-C
- Solubility in Methanol:almost transparency
Frequently Bought Together