Order Entry
Italy
ContactUsLinkComponent
2'-Methylacetoacetanilide ≥98% (by HPLC) for synthesis, Sigma-Aldrich®
Irritant
2'-Methylacetoacetanilide ≥98% (by HPLC) for synthesis, Sigma-Aldrich®
Catalog # 8.43847.0100
Supplier:  Merck
CAS Number:  
Irritant
2'-Methylacetoacetanilide ≥98% (by HPLC) for synthesis, Sigma-Aldrich®
Catalog # 8.43847.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :191.23 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:202-267-0
 :GHS07
Storage Temperature:Ambient
CAS Number:93-68-5
Formula:C₁₁H₁₃NO₂
MDL Number:MFCD00008782