2',5'-Dichloroacetanilide ≥98.0% (by total nitrogen basis)
Catalog # TCIAD0307-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:134.0 to 137.0 deg-C
Specification Test Results
About this item
:
:204.0500 g/mol
EINECS:220-062-4
CAS Number:2621-62-7
Formula:C₈H₇Cl₂NO
MDL Number:MFCD00018519