Order Entry
Italy
ContactUsLinkComponent
4,5-Diaminopyrimidine ≥98.0% (by GC, titration analysis)
Irritant
4,5-Diaminopyrimidine ≥98.0% (by GC, titration analysis)
Catalog # TCIAD3858-1G
Supplier:  TCI
CAS Number:  
undefined
Irritant
4,5-Diaminopyrimidine ≥98.0% (by GC, titration analysis)
Catalog # TCIAD3858-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Gray to Brown powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    201.0 to 207.0 deg-C

Specification Test Results

About this item

 :
 :110.1200 g/mol
Boiling Pt:229°C
EINECS:237-339-0
 :GHS07
CAS Number:13754-19-3
Formula:C₄H₆N₄
MDL Number:MFCD00006113