Order Entry
Italy
ContactUsLinkComponent
Lithium perfluorobutane-1-sulphonate ≥95.0% (by titrimetric analysis)
Lithium perfluorobutane-1-sulphonate ≥95.0% (by titrimetric analysis)
Catalog # TCIAN0710-25G
Supplier:  TCI
CAS Number:  
undefined
Lithium perfluorobutane-1-sulphonate ≥95.0% (by titrimetric analysis)
Catalog # TCIAN0710-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Ion exchange titration):
    min. 95.0 %
  • Solubility in Water:
    almost transparency

Specification Test Results

About this item

 :
 :306.0326 g/mol
CAS Number:131651-65-5
Formula:C₄F₉LiO₃S
MDL Number:MFCD03094160