

N-Methyl-bis-trifluoroacetamide ≥97.0% (by GC) trifluoroacylating agent
Supplier: TCI
Certificates
About this item
CAS Number:685-27-8
:
Formula:C₅H₃F₆NO₂
MDL Number:MFCD00000412
:223.07 g/mol
Flash Pt:45°C
EINECS:211-680-5
UN:2920
ADR:8,II
:GHS02
:GHS05
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 97.0 %
- Effect test of GC derivalization:Effective as derivatization agent N-Methylaniline
Specification Test Results
Frequently Bought Together