
Ethyl (ethoxymethylene)acetate cis- and trans-mixture ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1001-26-9
:
Formula:C₇H₁₂O₃
MDL Number:MFCD00009863
:144.17 g/mol
Flash Pt:76°C
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 98.0 % (total of isomer)
Specification Test Results
Frequently Bought Together