Order Entry
Italy
ContactUsLinkComponent
Tetrabutylammonium dihydrogentrifluoride ≥90.0% (by titrimetric analysis)
Corrosive
Irritant
Tetrabutylammonium dihydrogentrifluoride ≥90.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
undefined
Corrosive
Irritant
Tetrabutylammonium dihydrogentrifluoride ≥90.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Light orange to Yellow clear liquid
  • Purity (Neutralization titration):
    min. 90.0 %

Specification Test Results

Frequently Bought Together