
1-Butyl-1-methylpyrrolidinium bis(trifluoromethylsulphonyl)imide ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:223437-11-4
:
Formula:C₁₁H₂₀F₆N₂O₄S₂
MDL Number:MFCD07784447
:422.41 g/mol
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (Ion exchange titration):min. 98.0 %
Specification Test Results
Frequently Bought Together