
1,3-Cyclohexanedione ≥98.0% (by GC, titration analysis) for HPLC labeling
Catalog # TCIAA5581-5G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:104.0 to 108.0 deg-C
- Solubility in Water:almost transparency
Specification Test Results
About this item
:
:112.13 g/mol
Merck Index:14,03178
Boiling Pt:235 °C (1013 hPa)
Melting Pt:105 °C
Density:1,25 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:207-980-0
:GHS07
CAS Number:504-02-9
Formula:C₆H₈O₂
MDL Number:MFCD00001585