
5,7-Dichloro-8-quinolinol ≥98.0% (by GC, titration analysis)
Catalog # TCIAD0412-5G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:179.0 to 183.0 deg-C
- Solubility in hot Acetone:almost transparency
Specification Test Results
About this item
:
:214.05 g/mol
Merck Index:14,02175
Boiling Pt:355 °C (1013 hPa)
Melting Pt:183 °C
EINECS:212-258-3
:GHS07
CAS Number:773-76-2
Formula:C₉H₅Cl₂NO
MDL Number:MFCD00006786