
α,α,α,α',α',α'-Hexafluoro-5-nitroxylene ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:328-75-6
:
Formula:C₈H₃F₆NO₂
MDL Number:MFCD00000384
:259.11 g/mol
Boiling Pt:72°C
Density:1,53 g/cm³ (20 °C)
Flash Pt:77°C
EINECS:206-336-6
:GHS07
Specification Test Results
- Appearance:Light yellow to Yellow to Orange clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together