



Benzyltrimethylammonium hydroxide 40% in methanol
Supplier: TCI
Certificates
About this item
CAS Number:100-85-6
:
Formula:C₁₀H₁₇NO
MDL Number:MFCD00008281
:167.25
Flash Pt:11°C
EINECS:202-895-5
UN:2924
ADR:3,II
:GHS02
:GHS05
:GHS07
:GHS08
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Concentration (Neutralization titration):40.0 to 44.0 w/w%
- Chloride:max. 1.5 %
Specification Test Results
Frequently Bought Together