
α,α,α-Trifluoro-3-iodotoluene ≥98.0% (by GC) stabilised
Catalog # TCIAI0385-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
About this item
:
:272.01 g/mol
Boiling Pt:183°C
Melting Pt:–8 °C
Density:1,887 g/cm³ (20 °C)
Flash Pt:70°C
EINECS:206-934-7
UN:3265
ADR:8,II
:Stabilised with copper chip
:GHS05
CAS Number:401-81-0
Formula:C₇H₄F₃I
MDL Number:MFCD00001049