Dimethylcyclopropane-1,1-dicarboxylate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:6914-71-2
:
Formula:C₇H₁₀O₄
MDL Number:MFCD00192079
:158.15 g/mol
Boiling Pt:198°C
Flash Pt:95°C
EINECS:414-240-2
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together