
4-(Diethylamino)benzaldehyde ≥99.0% (by GC, titration analysis)
: TCI
:120-21-8
:
:C₁₁H₁₅NO
:MFCD00003382
:177.25 g/mol
:174°C
:40 °C
:1,03 g/cm³ (20 °C)
:Refrigerator
:204-377-4
:6.1,III
:GHS07
- Appearance:Light yellow to Brown powder to crystal
- Purity (GC):min. 99.0 %
- Purity (Nonaqueous Titration):min. 99.0 %
- Melting point:39.0 to 43.0 deg-C
- Solubility in Methanol:almost transparency
Frequently Bought Together