trans-Dichloroethylene ≥98.0% (by GC) stabilised
: TCI
: (E)-1,2-Dichloroethene, trans-Dichloroethene, (E)-1,2-Dichloroethylene, trans-1,2-Dichloroethylene
:156-60-5
:
:C₂H₂Cl₂
:MFCD00062942
:48°C
:Stabilised with 4-methoxyphenol (MEHQ)
:96.94 g/mol
:-49
:2°C
:3 II
:14,00092
:205-860-2
:1150
:GHS02
:GHS07
:GHS09
- Environmentally Preferable:
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Frequently Bought Together