



n-Octane ≥98.5% (by GC) for spectrophotometry
TCIAO0118-100ML
: TCI
:
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
- Appearance:Colorless clear liquid
- Purity (GC):min. 98.5 %
- Absorbance (210nm):max. 0.500
- Absorbance (220nm):max. 0.150
- Absorbance (230nm):max. 0.050
:
:114.23 g/mol
:14,06749
:125°C
:-57°C
:–57°C
:0,703 g/cm³ (20 °C)
:14°C
:Ambient
:203-892-1
:1262
:3,II
:GHS02
:GHS07
:GHS08
:GHS09
:111-65-9
:H₃C(CH₂)₆CH₃
:MFCD00009556