n-Nonane ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:111-84-2
:
Formula:H₃C(CH₂)₇CH₃
MDL Number:MFCD00009574
Boiling Pt:150°C
Melting Pt:–51°C
Density:0,718 g/cm³ (20 °C)
Flash Pt:31°C
Storage Temperature:Ambient
EINECS:203-913-4
ADR:3,III
:128.26 g/mol
UN:1920
:GHS02
:GHS07
:GHS08
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together