Order Entry
Italy
ContactUsLinkComponent
n-Heptane ≥99.0% (by GC) sequencing solvent
Flammable
Irritant
Health
Environment
n-Heptane ≥99.0% (by GC) sequencing solvent
Catalog # TCIAQ0037-100ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
Irritant
Health
Environment
n-Heptane ≥99.0% (by GC) sequencing solvent
Catalog # TCIAQ0037-100ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Colorless clear liquid
  • Purity (GC):
    min. 99.0 %
  • Density (20deg.C):
    0.6820 to 0.6860 g/mL
  • Absorbance (210nm):
    max. 0.350
  • Absorbance (220nm):
    max. 0.150

Specification Test Results

About this item

 :
 :100.20 g/mol
Merck Index:14,04659
Boiling Pt:98°C
Melting Pt:–91°C
Density:0,6838 g/cm³ (20 °C)
Flash Pt:–2°C
Storage Temperature:Ambient
EINECS:205-563-8
UN:1206
ADR:3,II
 :GHS02
 :GHS07
 :GHS08
 :GHS09
CAS Number:142-82-5
Formula:H₃C(CH₂)₅CH₃
MDL Number:MFCD00009544