



n-Heptane ≥99.0% (by GC) for spectrophotometry
Catalog # TCIAH0027-100ML
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Appearance:Colorless clear liquid
- Purity (GC):min. 99.0 %
- Density (20deg.C):0.6820 to 0.6860 g/mL
- Absorbance (210nm):max. 0.350
- Absorbance (220nm):max. 0.150
Specification Test Results
About this item
:
:100.20 g/mol
Merck Index:14,04659
Boiling Pt:98°C
Melting Pt:–91°C
Density:0,6838 g/cm³ (20 °C)
Flash Pt:–2°C
Storage Temperature:Ambient
EINECS:205-563-8
UN:1206
ADR:3,II
:GHS02
:GHS07
:GHS08
:GHS09
CAS Number:142-82-5
Formula:H₃C(CH₂)₅CH₃
MDL Number:MFCD00009544