
1,5-Diiodopentane ≥98.0% (by GC) stabilised
TCIAD2110-25G
: TCI
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Appearance:Light yellow to Yellow to Orange clear liquid
- Purity (GC):min. 98.0 %
:
:323.94 g/mol
:102°C
:9 °C
:2,195 g/cm³ (20 °C)
:Stabilised with copper chip
:211-054-1
:GHS07
:628-77-3
:C₅H₁₀I₂
:MFCD00001101