
4-Bromostyrene ≥95.0% (by GC) stabilised
Supplier: TCI
Certificates
About this item
CAS Number:2039-82-9
:
Formula:C₈H₇Br
MDL Number:MFCD00000110
:183.05 g/mol
Boiling Pt:81°C
Melting Pt:4.5 °C
Density:1,5991 g/cm³ (20 °C)
Flash Pt:75°C
:Stabilised with 4-tert-butylpyrocatechol (TBC)
EINECS:218-022-6
:GHS07
Storage Temperature:Freezer
Specification Test Results
- Appearance:Colorless to Yellow to Orange clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
Frequently Bought Together