
3-Bromostyrene ≥97.0% (by GC) stabilised
: TCI
:2039-86-3
:
:C₈H₇Br
:MFCD00000088
:183.05 g/mol
:94°C
:1,4059 g/cm³ (20 °C)
:67°C
:Stabilised with 4-tert-butylpyrocatechol (TBC)
:218-025-2
:GHS07
:Refrigerator
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 97.0 %
Frequently Bought Together