L(-)-Sorbose ≥98.0% (by HPLC)
TCIAS0066-25G
: TCI
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:155.0 to 165.0 deg-C
- Specific rotation [a]20/D:-42.0 to -44.0 deg (C=1, H2O)
- Solubility in Water:almost transparency
:
:180.16 g/mol
:14,08726
:380 °C (1013 hPa)
:158…160 °C
:Ambient
:201-771-8
:87-79-6
:C₆H₁₂O₆
:MFCD00151097