Order Entry
Italy
ContactUsLinkComponent
Octadecyltrichlorosilane ≥85.0% (by GC)
Corrosive
Octadecyltrichlorosilane ≥85.0% (by GC)
Catalog # TCIAO0079-25G
Supplier:  TCI
CAS Number:  
undefined
Corrosive
Octadecyltrichlorosilane ≥85.0% (by GC)
Catalog # TCIAO0079-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White or Colorless to Light yellow powder to lump to clear liquid
  • Purity (GC):
    min. 85.0 %

Specification Test Results

About this item

 :
 :387.94 g/mol
Boiling Pt:162°C
Melting Pt:22 °C
Density:0,98 g/cm³ (20 °C)
Flash Pt:191°C
EINECS:203-930-7
UN:1800
ADR:8,II
 :GHS05
CAS Number:112-04-9
Formula:C₁₈H₃₇Cl₃Si
MDL Number:MFCD00000484