
4-Bromo-α,α,α,2,3,5,6-heptafluorotoluene ≥99.0% (by GC)
TCIAT1541-10G
: TCI
:
- Pack type:Glass bottle
- Pk:10 g
- Seq:3236
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 99.0 %
:
:296.97 g/mol
:153°C
:1,92 g/cm³ (20 °C)
:62°C
:GHS07
:17823-46-0
:C₇BrF₇
:MFCD00075240