
(±)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol ≥97.0% (by GC)
: TCI
:100-79-8
:
:C₆H₁₂O₃
:MFCD00063238
:132.16 g/mol
:14,05213
:80°C
:1,04 g/cm³ (20 °C)
:202-888-7
:GHS07
:Ambient
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 97.0 %
Frequently Bought Together