Order Entry
Italy
ContactUsLinkComponent
Tetrapropylammonium hydroxide ∼40% in water
 
Tetrapropylammonium hydroxide ∼40% in water
  TCIAT1565-25G
 :  TCI
 :  
undefined
 
Tetrapropylammonium hydroxide ∼40% in water
  TCIAT1565-25G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Concentration (Neutralization titration):
    40.0 to 44.0 w/w%
  • Bromide:
    max. 0.01 %

 

 

 :
 :203.37 g/mol
 :Refrigerator
 :224-800-6
 :3267
 :8,II
 :GHS05
 :4499-86-9
 :C₁₂H₂₉NO
 :MFCD00009360