

Trimethylsilyl trifluoromethanesulphonate ≥98.0% (by titrimetric analysis) trimethylsilylating agent
Supplier: TCI
Certificates
About this item
CAS Number:27607-77-8
:
Formula:C₄H₉F₃O₃SSi
MDL Number:MFCD00000406
:222.26 g/mol
Merck Index:14,09719
Boiling Pt:140°C
Density:1,225 g/cm³ (20 °C)
Flash Pt:25°C
EINECS:248-565-4
UN:2920
ADR:8,II
:GHS02
:GHS05
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together