
Tetraethylammonium hydroxide 10% in water
Supplier: TCI
Certificates
About this item
CAS Number:77-98-5
:
Formula:C₈H₂₁NO
MDL Number:MFCD00009024
:147.26 g/mol
Merck Index:14,09201
EINECS:201-073-3
UN:3267
ADR:8,II
:GHS05
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Concentration (Neutralization titration):10.0 to 11.0 w/w%
Specification Test Results
Frequently Bought Together