Order Entry
Italy
ContactUsLinkComponent
trans-1,1-Diethoxy-2-hexene ≥95.0% (by GC)
Irritant
trans-1,1-Diethoxy-2-hexene ≥95.0% (by GC)
Catalog # TCIAH0575-25ML
Supplier:  TCI
CAS Number:  
undefined
Irritant
trans-1,1-Diethoxy-2-hexene ≥95.0% (by GC)
Catalog # TCIAH0575-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 95.0 %

Specification Test Results

About this item

 :
 :172.27 g/mol
Flash Pt:65°C
EINECS:266-989-8
 :GHS07
CAS Number:67746-30-9
Formula:C₁₀H₂₀O₂
MDL Number:MFCD00009472