Order Entry
Italy
ContactUsLinkComponent
 
 
2,5-Dimethoxyphenylboronic acid (contains varying amounts of Anhydride)
 :  TCI
 
undefined
 
2,5-Dimethoxyphenylboronic acid (contains varying amounts of Anhydride)
 :  TCI

 

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (Neutralization titration):
    97.0 to 110.0 %

 

Frequently Bought Together