EDTA disodium salt dihydrate ≥99.0% (by titrimetric analysis) for biochemical research
Supplier: TCI
Certificates
About this item
CAS Number:6381-92-6
:
Formula:C₁₀H₁₄N₂Na₂O₈·2H₂O
MDL Number:MFCD00150037
:372.24 g/mol
Merck Index:13,03543
Melting Pt:252°C
Storage Temperature:Ambient
EINECS:205-358-3
Specification Test Results
- Appearance:White powder to crystaline
- Purity (Chelometric Titration):min. 99.0 %
- Enzyme inhibition test:to pass test
Specification Test Results
Frequently Bought Together