
1-Methylcyclopropanecarboxylic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:6914-76-7
:
Formula:C₅H₈O₂
MDL Number:MFCD00001290
:100.12 g/mol
EINECS:230-020-7
UN:3261
ADR:8,III
:GHS05
Specification Test Results
- Appearance:White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together