Span® 40
Nome del fornitore: TCI
About this item
Synonyms: Sorbitan monopalmitate, Arlacel 40
- CAS Number:26266-57-9
- Formula:C₂₂H₄₂O₆
- MDL Number:MFCD00080946
- Molecular Weight:402.57 g/mol
- Boiling Pt:? 100 °C (1013 hPa)
- Density:1 g/cm³ (20 °C)
- Flash Pt:≥ 149 °C
- Storage Temperature:Ambient
- EINECS:247-568-8
2 Options Available Below
Product Details & Documents
Synonyms: Sorbitan monopalmitate, Arlacel 40
- CAS Number:26266-57-9
- Formula:C₂₂H₄₂O₆
- MDL Number:MFCD00080946
- Molecular Weight:402.57 g/mol
- Boiling Pt:? 100 °C (1013 hPa)
- Density:1 g/cm³ (20 °C)
- Flash Pt:≥ 149 °C
- Storage Temperature:Ambient
- EINECS:247-568-8
Specifications for Product Family
Specification Test Results
- Appearance:Light yellow to Brown to Dark green pellet
- Melting point:45.0 to 48.0 deg-C
- Saponification Value:135.0 to 158.0



