Order Entry
Italy
ContactUsLinkComponent
2,4-Dimethylphenylboronic acid (contains varying amounts of Anhydride)
Irritant
2,4-Dimethylphenylboronic acid (contains varying amounts of Anhydride)
Supplier:  TCI
Certificates
undefined
Irritant
2,4-Dimethylphenylboronic acid (contains varying amounts of Anhydride)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (Neutralization titration):
    97.0 to 116.0 %

Specification Test Results

Related Information

Frequently Bought Together