Order Entry
Italy
ContactUsLinkComponent
Imidazole ≥99.0% (by GC, titration analysis) for fluorimetric analysis
Corrosive
Irritant
Health
Imidazole ≥99.0% (by GC, titration analysis) for fluorimetric analysis
Catalog # TCIAI0290-25G
Supplier:  TCI
CAS Number:  
undefined
Corrosive
Irritant
Health
Imidazole ≥99.0% (by GC, titration analysis) for fluorimetric analysis
Catalog # TCIAI0290-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White powder to crystal
  • Purity (GC):
    min. 99.0 %
  • Purity (Nonaqueous Titration):
    min. 99.0 %
  • Melting point:
    88.0 to 91.0 deg-C
  • Intensity of fluorescence:
    to pass test

Specification Test Results

About this item

 :
Merck Index:14,04912
Boiling Pt:256°C
Melting Pt:89…90 °C
Density:1,18 g/cm³ (20 °C)
Flash Pt:135 °C
Storage Temperature:Ambient
EINECS:206-019-2
UN:2923
ADR:8,III
 :68.08 g/mol
 :GHS05
 :GHS07
 :GHS08
CAS Number:288-32-4
Formula:C₃H₄N₂
MDL Number:MFCD00005183