


2-Mercaptoethanol ≥99.0% (by GC) for electrophoresis
Supplier: TCI
Certificates
About this item
CAS Number:60-24-2
:
Formula:HSCH₂CH₂OH
MDL Number:MFCD00004890
Merck Index:14,05869
Boiling Pt:158°C
Melting Pt:–100°C
Density:1,12 g/cm³ (20 °C)
Flash Pt:74°C
Storage Temperature:Ambient
UN:2966
ADR:6.1,II
:78.14 g/mol
EINECS:200-464-6
:GHS06
:GHS08
:GHS09
Specification Test Results
- Appearance:Colorless clear liquid
- Purity (GC):min. 99.0 %
- Refractive index n20/D:1.5000 to 1.5040
- Functionality test:to pass test
Specification Test Results
Frequently Bought Together