Order Entry
Italy
Orders LinkContactUsLinkComponent
Span® 20

Span® 20

Supplier:   TCI
Span® 20
Supplier:  TCI
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  Sorbitan monododecanoate, Arlacel 20, Sorbitan monolaurate
  • CAS Number:1338-39-2
  • Formula:C₁₈H₃₄O₆
  • MDL Number:MFCD00005365
  • Molecular Weight:346.46 g/mol
  • Merck Index:14,08724
  • Density:1,05 g/cm³ (20 °C)
  • Flash Pt:204°C
  • Storage Temperature:Ambient
  • EINECS:215-663-3
Specifications for Product Family

Specification Test Results

  • Appearance:
    Light yellow to Brown clear liquid
  • Saponification Value:
    155.0 to 174.0
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 18,00 €
    18,00 €
    1 * 25 g
       18,00 €

  • Glass bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 33,00 €
    33,00 €
    1 * 500 g
       33,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.