
1,1-Dichloro-3,3-dimethylbutanone ≥98.0% (by GC)
Catalog # TCIAD2233-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:50.0 to 52.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
About this item
:
:169.05 g/mol
Boiling Pt:178°C
EINECS:245-111-7
UN:2811
ADR:6.1,III
:GHS06
CAS Number:22591-21-5
Formula:C₆H₁₀Cl₂O
MDL Number:MFCD00069423