Order Entry
Italy
Orders LinkContactUsLinkComponent
VWR®, Multi-sectored samplers
VWR®, Multi-sectored samplers
Supplier:   VWR Collection
VWR®, Multi-sectored samplers
Supplier:  VWR Collection
Certificates
VWR®, Multi-sectored samplers
VWR®, Multi-sectored samplers
6 Options Available Below
Product Details & Documents
Multi-sectored samplers for sampling powders, pellets and friable materials, which may become non homogeneous in bags at various levels. The models with separate sections are mainly used for chemical or physical analysis; models with communicating sections are used for microbiological analysis.
  • Penetrating tip in stainless steel for all models
Suitable for collecting cereals, grains, seeds and rice from trucks, bulk containers and silos - made from anodised aluminium or stainless steel.
Method of use: The T handle is turned to close the cells and the steel point is inserted through the bag wall or the commodity. When the sampler is deep enough to cover the different sectors, the handle is turned a half turn to allow samples to enter and then returned to the original position. The sampler is then removed and the different samples placed into containers for analysis.
Product Family Options
Product Information
  • Material
    Length
    Ø
    Capacity
    Availability
    Price
  • 300-0170
    Sampler with 3 separated sectors (378 g)
    Sampler with 3 separated sectors (378 g)
    Aluminium
    560 mm
    20 mm
    20 ml
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 340,00 €
    340,00 €
    1 * 1 pezzi
       340,00 €
  • 300-0168
    Sampler with 3 separated sectors (618 g)
    Sampler with 3 separated sectors (618 g)
    Aluminium
    860 mm
    20 mm
    25 ml
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 207,00 €
    207,00 €
    1 * 1 pezzi
       207,00 €
  • 300-0169
    Sampler with 3 comunicant sectors (296 g)
    Sampler with 3 comunicant sectors (296 g)
    Aluminium
    550 mm
    20 mm
    44 ml
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 208,00 €
    208,00 €
    1 * 1 pezzi
       208,00 €
  • 300-0167
    Sampler with 3 comunicant sectors (434 g)
    Sampler with 3 comunicant sectors (434 g)
    Aluminium
    860 mm
    20 mm
    73 ml
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi
  • 300-0292
    Sampler with 3 separated sectors (1011 g)
    Sampler with 3 separated sectors (1011 g)
    Stainless steel
    560 mm
    20 mm
    20 ml
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi
  • 300-0293
    Sampler with 3 separated sectors (1655 g)
    Sampler with 3 separated sectors (1655 g)
    Stainless steel
    860 mm
    20 mm
    20 ml
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi