Disinfectant, aerial, dry treatment of surfaces, 'Fumispore OPP'
A dry treatment which is effective in killing fungi, yeasts and bacteria with wide ranging applications in the food industry.
- Compliant with European Biocide Directive
- Easy to use, self contained, with no special equipment required
- Treatment takes a few hours and can easily be done overnight
- Activity based on orthophenylphenol
: Use biocides safely. Always read the label and product information before use.
- Catalog No:
- LBCBFSPOPPIT0400
- LBCBFSPOPPIT1000
- LBCBFSPOPPIT0040
- LBCBFSPOPPIT0200
- LBCBFSPOPPIT0020
- Type:
- FUMISPORE OPP
- FUMISPORE OPP
- FUMISPORE OPP
- FUMISPORE OPP
- FUMISPORE OPP
- Bactericidal activity:
- Listeria monocytogenes, E. coli, Pseudomonas aeruginosa, Salmonella enteritidis
- Listeria monocytogenes, E. coli, Pseudomonas aeruginosa, Salmonella enteritidis
- Listeria monocytogenes, E. coli, Pseudomonas aeruginosa, Salmonella enteritidis
- Listeria monocytogenes, E. coli, Pseudomonas aeruginosa, Salmonella enteritidis
- Listeria monocytogenes, E. coli, Pseudomonas aeruginosa, Salmonella enteritidis
- Fungicidal activity:
- Candida albicans, Penicillium chrysogenum, Penicillium cyclopium, Aspergillus fumigatus, Aspergillus niger, Aspergillus versicolor, Absidia corymbifera
- Candida albicans, Penicillium chrysogenum, Penicillium cyclopium, Aspergillus fumigatus, Aspergillus niger, Aspergillus versicolor, Absidia corymbifera
- Candida albicans, Penicillium chrysogenum, Penicillium cyclopium, Aspergillus fumigatus, Aspergillus niger, Aspergillus versicolor, Absidia corymbifera
- Candida albicans, Penicillium chrysogenum, Penicillium cyclopium, Aspergillus fumigatus, Aspergillus niger, Aspergillus versicolor, Absidia corymbifera
- Candida albicans, Penicillium chrysogenum, Penicillium cyclopium, Aspergillus fumigatus, Aspergillus niger, Aspergillus versicolor, Absidia corymbifera
- Sporicidal activity:
- Cladosporium cladosporoides
- Cladosporium cladosporoides
- Cladosporium cladosporoides
- Cladosporium cladosporoides
- Cladosporium cladosporoides
Frequently Bought Together