Culture media, ready to use, in plates
Supplier: PBI Brand
Certificates
About this item
Petri plates for environmental monitoring are designed for quality control of air contamination by the exposure method. Irradiated double or triple packing allows the insertion of the plates in the area to be monitored without any environmental contamination. By removing the most external protection before entering the white zone, it allows the material to enter the contaminated area. It is possible to request formulations and custom packaging as needed.
Frequently Bought Together