Order Entry
Italy
ContactUsLinkComponent
Microorganisms, KWIK-STIK™ Plus
Microorganisms, KWIK-STIK™ Plus
Catalog # 0258X
Supplier:  MICROBIOLOGICS
CAS Number:  
Microorganisms, KWIK-STIK™ Plus
Catalog # 0258X
Supplier:  MICROBIOLOGICS
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Pk:
    1
  • Description:
    Bacillus pumilus ATCC® 14884™
  • Seq:
    04
Specifications
About this item
Two passage microorganisms. KWIK-STIK™ Plus microorganisms are designed to meet the needs of laboratories requiring QC microorganisms that are a limited number of passages from the reference culture. Each KWIK-STIK™ Plus microorganism preparation is just two passages from the reference culture, allowing for more subcultures. Conveniently packaged in a KWIK-STIK™ device featuring a lyophilized microorganism pellet, inoculating swab and reservoir of hydrating fluid, KWIK-STIK™ Plus is ready to use and affordable. For added convenience, a peel-off Certificate of Assay is included for quality control documentation. The ATCC® licensed derivative emblem, the ATCC® licensed derivative word mark and the ATCC® catalog marks are trademarks of ATCC®. Microbiologics, Inc. is licensed to use these trademarks and to sell products derived from ATCC® cultures.
  • Two passages from reference culture
  • Microorganism, hydrating fluid and inoculating swab all in one self-contained device
  • Product warranty
Packaging: KWIK-STIK™ Plus includes five STIKS of a single strain.
Attention: Other microorganisms are available in the same format. Please contact Avantor to get more information.